Merck
  • Identification and characterization of PKCγ, a kinase associated with SCA14, as an amyloidogenic protein.
Identification and characterization of PKCγ, a kinase associated with SCA14, as an amyloidogenic protein.
Human molecular genetics (2014-09-14)
Hideyuki Takahashi, Naoko Adachi, Toshihiko Shirafuji, Sally Danno, Takehiko Ueyama, Michele Vendruscolo, Anton N Shuvaev, Takuya Sugimoto, Takahiro Seki, Daizo Hamada, Kazuhiro Irie, Hirokazu Hirai, Norio Sakai, Naoaki Saito
ABSTRACT
Amyloid assemblies are associated with a wide range of human disorders, including Alzheimer's and Parkinson's diseases. Here, we identify protein kinase C (PKC) γ, a serine/threonine kinase mutated in the neurodegenerative disease spinocerebellar ataxia type 14 (SCA14), as a novel amyloidogenic protein with no previously characterized amyloid-prone domains. We found that overexpression of PKCγ in cultured cells, as well as in vitro incubation of PKCγ without heat or chemical denaturants, causes amyloid-like fibril formation of this protein. We also observed that SCA14-associated mutations in PKCγ accelerate the amyloid-like fibril formation both in cultured cells and in vitro. We show that the C1A and kinase domains of PKCγ are involved in its soluble dimer and aggregate formation and that SCA14-associated mutations in the C1 domain cause its misfolding and aggregation. Furthermore, long-term time-lapse imaging indicates that aggregates of mutant PKCγ are highly toxic to neuronal cells. Based on these findings, we propose that PKCγ could form amyloid-like fibrils in physiological and/or pathophysiological conditions such as SCA14. More generally, our results provide novel insights into the mechanism of amyloid-like fibril formation by multi-domain proteins.