Merck
  • Expression of podoplanin/D2-40 in pericryptal stromal cells in superficial colorectal epithelial neoplasia.
Expression of podoplanin/D2-40 in pericryptal stromal cells in superficial colorectal epithelial neoplasia.
Medical molecular morphology (2013-01-12)
Hirofumi Nakayama, Hideaki Enzan, Wataru Yasui
ABSTRACT
The aim of this study is to investigate the distribution and roles of podoplanin/D2-40-positive pericryptal stromal cells in superficial colorectal epithelial neoplasia. A total of 105 superficial colorectal epithelial tumors were examined: 65 tubular/tubulovillous adenomas, 32 adenocarcinomas in situ, and 8 submucosally invasive adenocarcinomas. Immunohistochemical analysis was performed using the monoclonal antibody to podoplanin/clone D2-40, which is reactive in both lymphatic endothelial cells and activated stromal cells, but negative in vascular endothelial cells. We found 50 (78xa0%) of 65 tubular/tubulovillous adenomas, 30 (94xa0%) of 32 adenocarcinomas in situ, and all 8 (100xa0%) submucosally invasive adenocarcinomas had podoplanin/D2-40-positive pericryptal stromal cells, whereas all normal colorectal mucosae had no podoplanin/D2-40-positive pericryptal stromal cells. The presence of podoplanin/D2-40-positive pericryptal stromal cells is associated with epithelial tumorigenesis in the colorectum.