Merck
  • Correlation Between E-cadherin Immunoexpression and Efficacy of First Line Platinum-Based Chemotherapy in Advanced High Grade Serous Ovarian Cancer.
Correlation Between E-cadherin Immunoexpression and Efficacy of First Line Platinum-Based Chemotherapy in Advanced High Grade Serous Ovarian Cancer.
Pathology oncology research : POR (2014-08-12)
Branka Petrić Miše, Vesna Dobrić Telesmanić, Snježana Tomić, Dinka Šundov, Vesna Čapkun, Eduard Vrdoljak
ABSTRACT
To analyze correlation between immunoexpression of E-cadherin and efficacy of first line platinum-based chemotherapy in patients with advanced-stage high-grade serous ovarian carcinoma. The expression of E-cadherin was analyzed immunohistochemically in formalin-fixed, paraffin-embedded samples from 98 patients with advanced-stage high-grade serous ovarian cancer and related to clinical features (stage according to the International Federation of Gynecology and Obstetrics (FIGO) and residual tumors after initial cytoreductive surgery), response to platinum-based chemotherapy (according to Response Evaluation Criteria in Solid tumors (RECIST 1.1 criteria)), platinum sensitivity (according to platinum free interval (PFI) as platinum-refractory, platinum-resistant and platinum-sensitive) and patients progression free survival (PFS) and overall survival (OS). E-cadherin immunostaining was positive in 74 and negative in 24 serous ovarian carcinomas. E-cadherin immunoreactivity was not associated with FIGO stage, residual tumor after initial cytoreductive surgery and number of chemotherapy cycles. Positive E-cadherin expression predict significantly better response to first line platinum-based chemotherapy (p < 0.001) and platinum sensitivity (p < 0.001). Moreover, positive E-cadherin expression predict significantly longer PFS (p < 0.001) and OS (p < 0.001). The multivariate analysis for OS showed that positive E-cadherin expression is predictor to platinum sensitivity (p < 0.001) and longer OS (p = 0.01). Positive E-cadherin expression seems to be a predictor of better response to first line platinum-based chemotherapy, platinum sensitivity and favorable clinical outcome in patients with advanced-stage serous ovarian cancer. Negative E-cadherin expression was shown to be significant, independent predictor of poorer PFS and OS. E-cadherin as a marker has predictive and prognostic value.