Merck
  • Direct analysis in real time - high resolution mass spectrometry (DART-HRMS): a high throughput strategy for identification and quantification of anabolic steroid esters.
Direct analysis in real time - high resolution mass spectrometry (DART-HRMS): a high throughput strategy for identification and quantification of anabolic steroid esters.
Drug testing and analysis (2014-09-30)
Mickael Doué, Gaud Dervilly-Pinel, Karinne Pouponneau, Fabrice Monteau, Bruno Le Bizec
ABSTRACT
High throughput screening is essential for doping, forensic, and food safety laboratories. While hyphenated chromatography-mass spectrometry (MS) remains the approach of choice, recent ambient MS techniques, such as direct analysis in real time (DART), offer more rapid and more versatile strategies and thus gain in popularity. In this study, the potential of DART hyphenated with Orbitrap-MS for fast identification and quantification of 21 anabolic steroid esters has been evaluated. Direct analysis in high resolution scan mode allowed steroid esters screening by accurate mass measurement (Resolution = 60 000 and mass error < 3 ppm). Steroid esters identification was further supported by collision-induced dissociation (CID) experiments through the generation of two additional ions. Moreover, the use of labelled internal standards allowed quantitative data to be recovered based on isotopic dilution approach. Linearity (R(2)  > 0.99), dynamic range (from 1 to 1000 ng mL(-1) ), bias (<10%), sensitivity (1 ng mL(-1) ), repeatability and reproducibility (RSD < 20%) were evaluated as similar to those obtained with hyphenated chromatography-mass spectrometry techniques. This innovative high throughput approach was successfully applied for the characterization of oily commercial preparations, and thus fits the needs of the competent authorities in the fight against forbidden or counterfeited substances.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Testosterone propionate, solid
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg