Merck
  • Ticarcillin: a review of its pharmacological properties and therapeutic efficacy.
Ticarcillin: a review of its pharmacological properties and therapeutic efficacy.
Drugs (1980-11-01)
R N Brogden, R C Heel, T M Speight, G S Avery
ABSTRACT
Ticarcillin is a semisynthetic penicillin for parenteral administration. The antibacterial activity of ticarcillin is similar to that of carbenicillin except that it is two to four times more active in vitro against Pseudomonas aeruginosa, generally less active against Gram-positive cocci and more active against most Gram-negative bacilli. As the pharmacokinetics of ticarcillin and carbenicillin are also similar, ticarcillin should theoretically be clinically effective when administered at a lower dosage than carbenicillin. There is some evidence that ticarcillin is comparable in efficacy with carbenicillin when given in half to two-thirds the dosage, when the drugs are given in combination with an aminoglycoside and in clinical situations where these drugs are agents of choice. Ticarcillin has been used successfully in the treatment of complicated urinary tract infection, pulmonary infection in cystic fibrosis and bacteraemia and is effective when combined with an aminoglycoside in severe infections in patients with granulocytopenia. The efficacy in anaerobic infections is at present poorly documented, although preliminary results are promising. Tolerability has generally been good with hypokalaemia being the most frequently reported side effect. At the dosages used, bleeding and fluid overload have seldom occurred.