Merck
  • A synthetic antagonist for the peroxisome proliferator-activated receptor gamma inhibits adipocyte differentiation.
A synthetic antagonist for the peroxisome proliferator-activated receptor gamma inhibits adipocyte differentiation.
The Journal of biological chemistry (2000-01-15)
H M Wright, C B Clish, T Mikami, S Hauser, K Yanagi, R Hiramatsu, C N Serhan, B M Spiegelman
ABSTRACT
While searching for natural ligands for the peroxisome proliferator-activated receptor (PPAR) gamma, we identified a synthetic compound that binds to this receptor. Bisphenol A diglycidyl ether (BADGE) is a ligand for PPARgamma with a K(d(app)) of 100 microM. This compound has no apparent ability to activate the transcriptional activity of PPARgamma; however, BADGE can antagonize the ability of agonist ligands such as rosiglitazone to activate the transcriptional and adipogenic action of this receptor. BADGE also specifically blocks the ability of natural adipogenic cell lines such as 3T3-L1 and 3T3-F442A cells to undergo hormone-mediated cell differentiation. These results provide the first pharmacological evidence that PPARgamma activity is required for the hormonally induced differentiation of adipogenic cells.