Merck
Chemical constituents from the fungus Chaetomium cupreum RY202.
Archives of pharmacal research (2014-06-11)
Natcha Panthama, Somdej Kanokmedhakul, Kwanjai Kanokmedhakul, Kasem Soytong
ABSTRACT
Two new angular types of azaphilones, isochromophilonol (1) and ochrephilonol (2), together with ten known compounds (3-12), were isolated from Chaetomium cupreum RY202. Their structures were established on the basis of spectroscopic data and the absolute configurations of 1 and 3 were determined by the modified Mosher's method. Compounds 1, 2 and 4 showed moderated cytotoxicity against KB and NCI-H187 cell lines (IC50 9.63-32.42 µg/mL).
Sigma-Aldrich
p-Anisaldehyde, 98%
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG