Merck
  • Transcription factor KLF4 regulates microRNA-544 that targets YWHAZ in cervical cancer.
Transcription factor KLF4 regulates microRNA-544 that targets YWHAZ in cervical cancer.
American journal of cancer research (2015-08-14)
Langyong Mao, Yan Zhang, Xiaolong Deng, Wenjuan Mo, Yao Yu, Hong Lu
ABSTRACT
The deregulation of microRNAs has been demonstrated in various tumor processes. Here, we report that microRNA-544 (miR-544) is decreased in cervical cancer tissues compared with normal cervical tissues. To identify the mechanisms involved in miR-544 deregulation, we studied the regulation of miR-544 expression at the transcriptional level. We first identified the transcriptional start site of miR-544 by 5' rapid amplification of cDNA ends and subsequently determined the miR-544 promoter. We discovered that the transcription factor Krueppel-like factor 4 (KLF4) is involved in the transcriptional regulation of miR-544 through interaction with the miR-544 promoter. In addition, we found that miR-544 directly targets the YWHAZ oncogene and functions as a tumor suppressor in cervical cancer cells. miR-544 is involved in cell cycle regulation and suppresses cervical cancer cell proliferation, colony formation, migration and invasion in a manner associated with YWHAZ downregulation. In summary, our findings demonstrate that KLF4 upregulates miR-544 transcription by activating the miR-544 promoter and that miR-544 functions as a tumor suppressor by targeting YWHAZ. Therefore, miR-544 may be a potential novel therapeutic target and prognostic marker for cervical cancer.