- Graphite sulphate – a precursor to graphene.
Graphite sulphate – a precursor to graphene.
Chemical communications (Cambridge, England) (2015-01-22)
Siegfried Eigler
PMID25605427
ABSTRACT
Graphite sulphate is used as a precursor to graphene for the first time. The positively charged graphene layers react with water to yield a processable graphene derivative. The unprecedented low density of defects is determined to be 0.06% on average and may open the way for electronic applications.