Merck
  • Effect of opioid receptor subtype-selective agonists on purinergic and adrenergic components of neurogenic contractions of mouse vas deferens.
Effect of opioid receptor subtype-selective agonists on purinergic and adrenergic components of neurogenic contractions of mouse vas deferens.
British journal of pharmacology (1993-02-01)
B Driessen, R Bültmann, I von Kügelgen, K Starke
ABSTRACT
1. Effects of opioid agonists on the purinergic and adrenergic components of neurogenic contractions and in some experiments on transmitter overflow were studied in the mouse isolated vas deferens. 2. When the vas deferens was stimulated every 2 min by pairs of pulses 2 s apart in the presence of prazosin 0.3 microM (to isolate the purinergic component) or alpha,beta-methylene-ATP 3 microM (to isolate the adrenergic component), each pulse elicited a separate twitch. The opioid agonists [D-Ala2,N-Me-Phe4,Gly5-ol]enkephalin (DAMGO, mu-receptor-selective), [D-Pen2,D-Pen5]enkephalin (DPDPE, delta-selective) and trans-(+/-)-3,4-dichloro-N-methyl-N-[2-(1-pyrrolidinyl)-cyclohexyl]- benzeneacetamide (U-50488, kappa-selective) concentration-dependently reduced both purinergic and adrenergic contractions. For each agonist, maximal effects and concentrations causing half-maximal effects were very similar for inhibition of the purinergic component on the one hand and for inhibition of the adrenergic component on the other hand, although the adrenergic component was inhibited with a slight preference. Moreover, effects on contractions elicited by the first and the second pulse of the pairs were very similar. 3. When vasa deferentia preincubated with [3H]-noradrenaline were stimulated with trains of 100 pulses delivered at 20 Hz, morphine 10 microM reduced significantly both evoked tritium overflow and evoked contractions. Its effect was antagonized by naloxone. 4. It is concluded that, in contrast to drugs acting at some other presynaptic receptors, opioid mu-, delta- and kappa-agonists inhibit purinergic and adrenergic neurogenic contractions of the mouse vas deferens in a similar manner. In contrast to a previous report, no enhancement by morphine of the release of noradrenaline elicited by high frequency pulse trains was observed.