Merck
  • An antioxidative galactomannan-protein complex isolated from fermentation broth of a medicinal fungus Cs-HK1.
An antioxidative galactomannan-protein complex isolated from fermentation broth of a medicinal fungus Cs-HK1.
Carbohydrate polymers (2014-08-19)
Xia Chen, Zhong-Yang Ding, Wen-Qiang Wang, Ka-Chai Siu, Jian-Yong Wu
ABSTRACT
A protein-containing polysaccharide, EPS2BW, was fractionated from the exopolysaccharide (EPS) produced by a medicinal fungus Cordyceps sinensis (Cs-HK1). EPS2BW was mainly composed of galactomannan with about 16% (w/w) protein and 50 kDa average molecular weight. The galactomannan part consisted of mannose and galactose at 1.7:1.0 molar ratio, and the protein segments were composed of sixteen amino acids with 12.5% proline and 16.6% threonine (mol%) being the most abundant. Based on analytical results from NMR, methylation analysis, partial acid hydrolysis and GC-MS, the galactomannan structure was elucidated as a (1 → 2)-α-D-mannopyranosyl (Manp) backbone with O-6-linked galactopyranosyl (Galp) branches. EPS2BW exhibited a high antioxidant capacity in both chemical and cell culture assays, with a Trolox equivalent radical scavenging activity of 44.7 μmol Trolox/mg, a Fe(3+) reducing power of 38.9 μmol Fe(2+)/mg, and significant cytoprotective effect against H2O2-induced PC12 cell death at 50-250 μg/mL.
Sigma-Aldrich
Anthrone, ACS reagent, 97%