Merck
  • Functional analysis of novel genetic variation in the thyroid hormone activating type 2 deiodinase.
Functional analysis of novel genetic variation in the thyroid hormone activating type 2 deiodinase.
The Journal of clinical endocrinology and metabolism (2014-08-21)
Chantal Zevenbergen, Wim Klootwijk, Robin P Peeters, Marco Medici, Yolanda B de Rijke, Sylvia A Huisman, Henk Goeman, Erik Boot, Gerda de Kuijper, K Herman de Waal, Marcel E Meima, P Reed Larsen, Theo J Visser, W Edward Visser
ABSTRACT
Thyroid hormones (TH) are important for normal brain development and abnormal TH regulation in the brain results in neurocognitive impairments. The type 2 deiodinase (D2) is important for local TH control in the brain by generating the active hormone T3 from its precursor T4. Dysfunction of D2 likely results in a neurocognitive phenotype. No mutations in D2 have been reported yet. The objective of the study was to identify D2 mutations in patients with intellectual disability and to test their functional consequences. The patients were selected from the multicenter Thyroid Origin of Psychomotor Retardation study, which is a cohort of 946 subjects with unexplained intellectual disability. Based on characteristic serum TH values, the coding region of the DIO2 gene was sequenced in 387 patients. Functional consequences were assessed by in vitro D2 assays or intact cell metabolism studies using cells transfected with wild-type or mutant D2. Sequence analysis revealed two heterozygous mutations: c.11T>A (p.L4H) in three subjects and c.305C>T (p.T102I) in one subject. Sequence analysis of family members revealed several carriers, but no segregation was observed with thyroid parameters or neurocognitive phenotype. Extensive tests with different in vitro D2 assays did not show differences between wild-type and mutant D2. This study describes the identification and functional consequences of novel genetic variation in TH activating enzyme D2. Family studies and functional tests suggest that these variants do not underlie the neurocognitive impairment. Altogether our data provide evidence of the existence of rare but apparently harmless genetic variants of D2.
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%