Merck
  • Role of activin-A and myostatin and their signaling pathway in human myometrial and leiomyoma cell function.
Role of activin-A and myostatin and their signaling pathway in human myometrial and leiomyoma cell function.
The Journal of clinical endocrinology and metabolism (2014-03-13)
Md Soriful Islam, William H Catherino, Olga Protic, Milijana Janjusevic, Peter Clarke Gray, Stefano Raffaele Giannubilo, Andrea Ciavattini, Pasquale Lamanna, Andrea Luigi Tranquilli, Felice Petraglia, Mario Castellucci, Pasquapina Ciarmela
ABSTRACT
Uterine leiomyomas are highly prevalent benign tumors of premenopausal women and the most common indication for hysterectomy. However, the exact etiology of this tumor is not fully understood. The objective of the study was to evaluate the role of activin-A and myostatin and their signaling pathways in human myometrial and leiomyoma cells. This was a laboratory study. Myometrial and leiomyoma cells (primary and cell lines) were cultured in vitro. The study included premenopausal women who were admitted to the hospital for myomectomy or hysterectomy. Primary myometrial and leiomyoma cells and/or cell lines were treated with activin-A (4 nM) and myostatin (4 nM) for different days of interval (to measure proliferation rate) or 30 minutes (to measure signaling molecules) or 48 hours to measure proliferating markers, extracellular matrix mRNA, and/or protein expression by real-time PCR, Western blot, and/or immunocytochemistry. We found that activin-A and myostatin significantly reduce cell proliferation in primary myometrial cells but not in leiomyoma cells as measured by a CyQUANT cell proliferation assay kit. Reduced expression of proliferating cell nuclear antigen and Ki-67 were also observed in myometrial cells in response to activin-A and myostatin treatment. Activin-A also significantly increased mRNA expression of fibronectin, collagen1A1, and versican in primary leiomyoma cells. Finally, we found that activin-A and myostatin activate Smad-2/3 signaling but do not affect ERK or p38 signaling in both myometrial and leiomyoma cells. This study results suggest that activin-A and myostatin can exert antiproliferative and/or fibrotic effects on these cell types via Smad-2/3 signaling.