Merck
  • Asymmetric and symmetric dimethylarginine are associated with coronary artery lesions in Kawasaki disease.
Asymmetric and symmetric dimethylarginine are associated with coronary artery lesions in Kawasaki disease.
The Journal of pediatrics (2014-05-31)
Ying-Hsien Huang, You-Lin Tain, Chiu-Ping Lee, Ho-Chang Kuo
ABSTRACT
To determine whether 3 biomarkers, L-arginine, asymmetric dimethylarginine (ADMA), and symmetric dimethylarginine (SDMA), can predict outcomes in patients with Kawasaki disease (KD). Plasma levels of L-arginine, ADMA, and SDMA were measured in 39 patients with KD and 27 febrile control patients. Plasma L-arginine, ADMA, and SDMA levels were lower in patients with KD than in control patients before treatment with intravenous immunoglobulin (IVIG; P=.027, P<.001, and P<.001, respectively). After treatment with IVIG, L-arginine, ADMA, L-arginine/ADMA ratios, and arginine methylation ([ADMA+SDMA]/L-arginine) increased significantly (P<.001, P=.001, P=.014, and P=.001, respectively). Compared with control patients, persistent lower SDMA and higher ADMA/SDMA ratios existed in patients with KD. Furthermore, a lesser magnitude of change in terms of L-arginine and ADMA/SDMA ratios after IVIG treatment was associated with the formation of coronary dilation (P=.025, and .029, respectively). Levels of L-arginine, ADMA, and SDMA appear to be associated with KD. Lower L-arginine levels and ADMA/SDMA after treatment with IVIG was associated with coronary artery abnormalities patients with KD.