Merck
  • Physiologic and metabolic safety of butyrylcholinesterase gene therapy in mice.
Physiologic and metabolic safety of butyrylcholinesterase gene therapy in mice.
Vaccine (2014-06-04)
Vishakantha Murthy, Yang Gao, Liyi Geng, Nathan K LeBrasseur, Thomas A White, Robin J Parks, Stephen Brimijoin
ABSTRACT
In continuing efforts to develop gene transfer of human butyrylcholinesterase (BChE) as therapy for cocaine addiction, we conducted wide-ranging studies of physiological and metabolic safety. For that purpose, mice were given injections of adeno-associated virus (AAV) vector or helper-dependent adenoviral (hdAD) vector encoding human or mouse BChE mutated for optimal cocaine hydrolysis. Age-matched controls received saline or AAV-luciferase control vector. At times when transduced BChE was abundant, physiologic and metabolic parameters in conscious animals were evaluated by non-invasive Echo-MRI and an automated "Comprehensive Laboratory Animal Monitoring System" (CLAMS). Despite high vector doses (up to 10(13) particles per mouse) and high levels of transgene protein in the plasma (∼1500-fold above baseline), the CLAMS apparatus revealed no adverse physiologic or metabolic effects. Likewise, body composition determined by Echo-MRI, and glucose tolerance remained normal. A CLAMS study of vector-treated mice given 40 mg/kg cocaine showed none of the physiologic and metabolic fluctuations exhibited in controls. We conclude that neither the tested vectors nor great excesses of circulating BChE affect general physiology directly, while they protect mice from disturbance by cocaine. Hence, viral gene transfer of BChE appears benign and worth exploring as a therapy for cocaine abuse and possibly other disorders as well.