Merck
  • Isolation of the insecticidal components of Tagetes minuta (Compositae) against mosquito larvae and adults.
Isolation of the insecticidal components of Tagetes minuta (Compositae) against mosquito larvae and adults.
Journal of the American Mosquito Control Association (1995-09-01)
M J Perich, C Wells, W Bertsch, K E Tredway
ABSTRACT
Application of Tagetes minuta floral extract to silica gel column chromatography produced 2 fractions with the hydrogenate part 20-30 times more toxic to larvae and 12-13 times more toxic to adults of Aedes aegypti and Anopheles stephensi, respectively, than the oxygenate part. Further fractionation by column chromatography of the hydrogenate fraction produced 4 thiophenes, 5-(but-3-ene-1-ynyl)-2,2'-bithiophene, 5-(but-3-ene-1-ynyl)-5'-methyl-2,2'-bithiophene, 2,2',5',2"-terthiophene, and 5-methyl-2,2',5',2"-terthiophene. These compounds in Tagetes minuta are largely responsible for the toxicity exhibited against the tested mosquitoes.