Merck
  • TP53INP1 as new therapeutic target in castration-resistant prostate cancer.
TP53INP1 as new therapeutic target in castration-resistant prostate cancer.
The Prostate (2012-01-04)
Sophie Giusiano, Virginie Baylot, Claudia Andrieu, Ladan Fazli, Martin Gleave, Juan Lucio Iovanna, Colette Taranger-Charpin, Stéphane Garcia, Palma Rocchi
ABSTRACT
Prostate cancer (PC) is one of the most common malignancies in industrialized countries, and the second leading cause of cancer-related death in the United States. We recently showed that over-expression of tumor protein 53-induced nuclear protein 1 (TP53INP1), a cell stress response protein, is a worse prognostic factor in PC, particularly predictive of biological cancer relapse. Moreover, treatment of castration-sensitive (CS) LNCaP tumor cells with a TP53INP1 antisense oligonucleotide (TP53INP1 ASO) inhibits proliferation and induces apoptosis. The aim of this study was to investigate variations of TP53INP1 expression in PC during androgen withdrawal therapy and in castration-resistant prostate cancer (CRPC). Quantitative measurements of immunohistochemical expression of TP53INP1 using high-throughput densitometry, assessed on digitized microscopic tissue micro-array images were correlated with hormone therapy (HT) status in human PC. Northern blot analysis of TP53INP1 after castration was performed in LNCaP xenograft. Treatment of CR C4-2 tumor cells in vitro with TP53INP1 ASO was analyzed. We also analyzed the effect of TP53INP1 ASO treatment in vivo on tumor xenograft growth. TP53INP1 protein expression decreases during HT and increases after HT in human CRPC. TP53INP1 mRNA increases significantly in CR tumors of LNCaP xenograft. Moreover, treatment of CR C4-2 cells with TP53INP1 ASO downregulates TP53INP1 protein level, inhibits proliferation, and induces apoptosis. Finally, in vivo, TP53INP1 ASO treatment significantly inhibits the tumoral progression of CR C4-2 xenograft and enhances docetaxel cytotoxicity. These results suggest that TP53INP1 could be considered as a relevant-specific target for molecular therapy of CRPC.