Merck
  • Mutually exclusive FGFR2, HER2, and KRAS gene amplifications in gastric cancer revealed by multicolour FISH.
Mutually exclusive FGFR2, HER2, and KRAS gene amplifications in gastric cancer revealed by multicolour FISH.
Cancer letters (2014-08-03)
Kakoli Das, Bavani Gunasegaran, Iain Beehuat Tan, Niantao Deng, Kiat Hon Lim, Patrick Tan
ABSTRACT
Gastric cancer (GC) is a major cause of global cancer mortality. Previous genomic studies have reported that several RTK-RAS pathway components are amplified in GC, with individual tumours often amplifying one component and not others ("mutual exclusivity"). Here, we sought to validate these findings for three RTK/RAS components (FGFR2, HER2, KRAS) using fluorescence in situ hybridisation (FISH) on a series of gastric tumours, cell lines and patient-derived xenografts. Applying dual-colour FISH on 137 gastric tumours (89 FFPE surgical resections and 48 diagnostic biopsies), we observed FGFR2 amplification in 7.3% and HER2 amplification in 2.2% of GCs. GCs exhibiting FGFR2 amplification were associated with high tumour grade (p = 0.034). In FISH positive tumours, striking differences in copy number levels between cancer cells in the same tumour were observed, suggesting intra-tumour heterogeneity. Using a multicolour FISH assay allowing simultaneous detection of FGFR2, HER2, and KRAS amplifications, we confirmed that these components exhibited a mutually exclusive pattern of gene amplification across patients. The FISH data were also strongly correlated with Q-PCR levels and at the protein level by immunohistochemistry. Our data confirm that RTK/RAS components are mutually exclusively amplified in GC, and demonstrate the feasibility of identifying multiple aneuploidies using a single FISH assay. Application of this assay to GC samples, particularly diagnostic biopsies, may facilitate enrollment of GC patients into clinical trials evaluating RTK/RAS directed therapies. However, the presence of intra-tumour heterogeneity may require multiple biopsy samples to be obtained per patient before a definitive diagnosis can be attained.
Sigma-Aldrich
DAPI, for nucleic acid staining