Merck
  • Undifferentiated (embryonal) sarcoma of the liver in middle-aged adults: smooth muscle differentiation determined by immunohistochemistry and electron microscopy.
Undifferentiated (embryonal) sarcoma of the liver in middle-aged adults: smooth muscle differentiation determined by immunohistochemistry and electron microscopy.
Human pathology (2003-04-04)
Jun Nishio, Hiroshi Iwasaki, Naomi Sakashita, Seiji Haraoka, Teruto Isayama, Masatoshi Naito, Haruhiko Miyayama, Yuuya Yamashita, Masahiro Kikuchi
ABSTRACT
Undifferentiated (embryonal) sarcoma of the liver (UESL) is a rare pediatric liver malignancy that is extremely uncommon in middle-aged individuals. We studied 2 cases of UESL in middle-aged adults (1 case in a 49-year-old woman and the other in a 62-year-old man) by histology, immunohistochemistry, and electron microscopy to clarify the cellular characteristics of this peculiar tumor. One tumor showed a mixture of spindle cells, polygonal cells, and multinucleated giant cells within a myxoid matrix and also revealed focal areas of a storiform pattern in a metastatic lesion. The other tumor was composed mainly of anaplastic large cells admixed with few fibrous or spindle-shaped components and many multinucleated giant cells. In both cases, some tumor cells contained eosinophilic hyaline globules that were diastase resistant and periodic acid-Schiff positive. Immunohistochemically, the tumor cells showed positive staining for smooth muscle markers, such as desmin, alpha-smooth muscle actin, and muscle-specific actin, and also for histiocytic markers, such as alpha-1-antitrypsin, alpha-1-antichymotrypsin, and CD68. Electron microscope examination revealed thin myofilaments with focal densities and intermediate filaments in the cytoplasm of tumor cells. Our studies suggest that UESL exhibits at least a partial smooth muscle phenotype in middle-aged adults, and this specific differentiation may be more common in this age group than in children. Tumor cells of UESL with smooth muscle differentiation in middle-aged adults show phenotypic diversity comparable to those of malignant fibrous histiocytoma with myofibroblastic differentiation.