- Preparation and relevance of a cross-coupling product between sinapyl alcohol and sinapyl p-hydroxybenzoate.
Preparation and relevance of a cross-coupling product between sinapyl alcohol and sinapyl p-hydroxybenzoate.
Organic & biomolecular chemistry (2004-10-14)
Fachuang Lu, John Ralph, Kris Morreel, Eric Messens, Wout Boerjan
PMID15480449
ABSTRACT
Cross-coupling of sinapyl p-hydroxybenzoate and sinapyl alcohol produces an 8-8-cross-coupled product that is also detected in lignifying poplar tissues, implicating sinapyl p-hydroxybenzoate as a lignin precursor.