- A one-pot multicomponent coupling reaction for the stereocontrolled synthesis of (Z)-trisubstituted allylic alcohols.
A one-pot multicomponent coupling reaction for the stereocontrolled synthesis of (Z)-trisubstituted allylic alcohols.
Journal of the American Chemical Society (2004-03-25)
Young K Chen, Patrick J Walsh
PMID15038709
ABSTRACT
In this Communication, we outline a new one-pot, multicomponent coupling reaction that allows easy access to (Z)-trisubstituted allylic alcohols. Our strategy is based on E to Z isomerization of the 1-bromo-1-dialkylvinylborane upon reaction with dialkylzinc reagents, and subsequent transmetalation to give (Z)-trisubstituted vinylzinc species. In situ trapping of the reactive vinylzinc intermediates with aldehydes furnished a series of (Z)-trisubstituted allylic alcohols. This method represents a viable alternative to the Still-Gennari modification of the HWE olefination reaction, and it has the advantage that it allows coupling of larger fragments.