Merck
  • Purification, recovery, and laser-driven fluorination of silicon from dissolved and particulate silica for the measurement of natural stable isotope abundances.
Purification, recovery, and laser-driven fluorination of silicon from dissolved and particulate silica for the measurement of natural stable isotope abundances.
Analytical chemistry (1996-11-01)
C L De La Rocha, M A Brzezinski, M J Deniro
ABSTRACT
A procedure for the purification, recovery, and determination of isotopic abundances of silicon from biogenic and lithogenic particulate matter and dissolved silicic acid is reported. Purification involves the reaction of acid molybdate with dissolved silicon in natural waters or that produced by the dissolution of particulate silica by hydrofluoric acid. The resulting silicomolybdic acid is then quantitatively precipitated by reaction with triethylamine hydrochloride. The silicon is recovered as silicon dioxide through stepwise combustion of the dried precipitate. Fluorination of the product for isotopic analysis is accomplished by laser heating under pure fluorine generated by the decomposition of a fluorine-based salt. The resulting silicon tetrafluoride is separated from hydrogen fluoride and other fluorination byproducts cryogenically using a variable-temperature cold trap. Yields for silicon recovery are 99.9% for precipitation and greater than 95% for the purification/fluorination procedure. Reproducibility of the isotopic composition for pure quartz granules processed through the procedure is ±0.1‰ for δ(30)Si.
Sigma-Aldrich
Triethylamine hydrochloride, ≥99.0% (AT)