Merck
Octadecylsilane-modified silicas in the adsorption of toluene.
Analytical and bioanalytical chemistry (2008-05-28)
Rodrigo Brambilla, Carolina F Pinto, Márcia S L Miranda, João H Z dos Santos
ABSTRACT
A series of octadecylsilane-modified silicas were prepared by sol-gel and grafting methods. Carbon contents and octadecyl chain conformations were shown to depend on the preparative route. Grafting engenders a low carbon content and a liquid-like chain conformation, while the sol-gel method affords a much higher carbon content and a crystalline conformation. The relationships between the toluene adsorption of the hybrid silicas and their chain conformations, their carbon contents and their textural characteristics are discussed. These sorbents, when used in combination with ultraviolet diffuse reflectance spectroscopy (UV DRS), can be employed as a rapid screening method for detection of aromatic compounds in water and air environmental matrices.