Merck
  • Real time imaging of supramolecular assembly formation via programmed nucleolipid recognition.
Real time imaging of supramolecular assembly formation via programmed nucleolipid recognition.
Journal of the American Chemical Society (2008-10-15)
Louis Moreau, Michel Camplo, Michel Wathier, Nada Taib, Michel Laguerre, Isabelle Bestel, Mark W Grinstaff, Philippe Barthélémy
ABSTRACT
Supramolecular assembly formation resulting from molecular recognition between complementary nucleolipids has been visualized in real time at the micrometer scale.