Merck
  • An AB block copolymer of oligo(methyl methacrylate) and poly(acrylic acid) for micellar delivery of hydrophobic drugs.
An AB block copolymer of oligo(methyl methacrylate) and poly(acrylic acid) for micellar delivery of hydrophobic drugs.
Journal of controlled release : official journal of the Controlled Release Society (1998-08-01)
T Inoue, G Chen, K Nakamae, A S Hoffman
ABSTRACT
An AB block copolymer of oligo(methyl methacrylate) (oMMA) and poly(acrylic acid) (PAAc) has been synthesized. The block copolymer forms micelles in an aqueous medium, as confirmed by a fluorescence probe technique using pyrene. Doxorubicin hydrochloride was incorporated into the micelle and the release profile of doxorubicin hydrochloride was investigated. Slow and prolonged release of doxorubicin hydrochloride from the micelle was observed. The AB block copolymer micelle can be useful for prolonged mucosal drug delivery of hydrophobic drugs.
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid), average Mv ~450,000