Merck
  • A high molar extinction coefficient sensitizer for stable dye-sensitized solar cells.
A high molar extinction coefficient sensitizer for stable dye-sensitized solar cells.
Journal of the American Chemical Society (2005-01-20)
Peng Wang, Cédric Klein, Robin Humphry-Baker, Shaik M Zakeeruddin, Michael Grätzel
ABSTRACT
An amphiphilic heteroleptic polypyridyl ruthenium complex with a high molar extinction coefficient was synthesized and demonstrated as an efficient, thermostable sensitizer in nanocrystalline dye-sensitized solar cells.