Merck
  • Sequence and structure comparison suggest that methionine aminopeptidase, prolidase, aminopeptidase P, and creatinase share a common fold.
Sequence and structure comparison suggest that methionine aminopeptidase, prolidase, aminopeptidase P, and creatinase share a common fold.
Proceedings of the National Academy of Sciences of the United States of America (1994-03-29)
J F Bazan, L H Weaver, S L Roderick, R Huber, B W Matthews
ABSTRACT
Amino acid sequence comparison suggests that the structure of Escherichia coli methionine aminopeptidase (EC 3.4.11.18) and the C-terminal domain of Pseudomonas putida creatinase (EC 3.5.3.3) are related. A detailed comparison of the three-dimensional folds of the two enzymes confirms this homology: with an approximately 260-residue chain segment, 218 C alpha atoms of the structures superimpose within 2.5 A; only 41 of these overlapping positions (i.e., 19%) feature identical amino acids in the two protein chains. Notwithstanding this striking correspondence in structure, methionine aminopeptidase binds and is stimulated by Co2+, while creatinase is not a metal-dependent enzyme. Searches of protein data banks using sequence and structure-based profiles reveal other enzymes, including aminopeptidase P (EC 3.4.11.9), prolidase (EC 3.4.13.9), and agropine synthase, that likely share the same "pita-bread" fold common to creatinase and methionine aminopeptidase.