- Facile lewis acid catalyzed synthesis of C(4) symmetric resorcinarenes
Facile lewis acid catalyzed synthesis of C(4) symmetric resorcinarenes
Organic letters (2000-12-02)
McIldowie, Mocerino, Skelton, White
PMID11101440
ABSTRACT
The Lewis acid catalyzed condensation of 3-methoxyphenol with octanal produced the C(4) symmetric calix[4]resorcinarene 2, in high yield. Of the numerous stereo- and regioisomers possible, the rccc isomer with C(4) symmetry was the only product isolated (as a racemate). The structure has been established by single-crystal X-ray structure analysis.