Merck
Fragrance material review on isoborneol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
S P Bhatia, D McGinty, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of isoborneol when used as a fragrance ingredient is presented.
Sigma-Aldrich
Isoborneol, ≥95%, FG