Merck
  • Felodipine/metoprolol: a review of the fixed dose controlled release formulation in the management of essential hypertension.
Felodipine/metoprolol: a review of the fixed dose controlled release formulation in the management of essential hypertension.
Drugs (2000-03-16)
M Haria, G L Plosker, A Markham
ABSTRACT
The main objective of fixed dose combination therapy for hypertension is to improve blood pressure (BP) control with lower, better tolerated dosages of 2 antihypertensives rather than higher dosages of a single agent. Felodipine and metoprolol lower BP via different, but complementary, mechanisms and controlled release formulations of these 2 drugs are available as a fixed dose combination, felodipine/metoprolol. In clinical trials in patients with hypertension, felodipine/metoprolol was significantly more effective than placebo and the respective monotherapies administered at the same dosages. Mean BP was reduced to < 155/90 mm Hg in patients treated with combination therapy and controlled in approximately 70% of patients. In one study that titrated dosages to effect, fewer felodipine/metoprolol than felodipine or metoprolol monotherapy recipients required dosage increases to achieve BP control (45 vs 60 and 67%, respectively). Data from double blind comparative studies show that the antihypertensive efficacy of felodipine/metoprolol 5 to 10/50 to 100 mg/day is significantly greater than that of enalapril monotherapy or captopril plus hydrochlorothiazide and equivalent to nifedipine/atenolol and amlodipine. In comparisons with enalapril, fewer felodipine/metoprolol than enalapril recipients required dosage titration to achieve BP control. Compared with amlodipine, felodipine/metoprolol significantly reduced mean 24-hour average BP (8.9/5.5 vs 14.4/9.5 mm Hg after 6 weeks; p < 0.001). Both treatments preserved diurnal rhythm. Long term follow-up studies show that the antihypertensive effect of felodipine/metoprolol occurs mostly during the first month of treatment with small additional decreases in BP being observed in the second and third months, and a relatively constant effect thereafter. According to a validated questionnaire, quality of life was relatively similar during 12 weeks treatment with felodipine/metoprolol, enalapril or placebo. In a retrospective pharmacoeconomic analysis conducted in Sweden, felodipine/metoprolol was more cost effective than enalapril as initial treatment for hypertension. Peripheral oedema, headache and flushing were the most commonly reported adverse events with felodipine/metoprolol and felodipine monotherapy, whereas dizziness, fatigue, headache and respiratory infection were more frequent with metoprolol monotherapy. Dose-dependent adverse events such as oedema may occur less often in patients taking lower dosages in combination than in those taking higher dosages of felodipine monotherapy. Thus, patients with hypertension treated with felodipine/metoprolol experience greater control of BP, with less need for dosage titration, than those treated with felodipine, metoprolol or enalapril monotherapy. Importantly this greater efficacy does not appear to be associated with a higher incidence of adverse events relative to monotherapy. Additionally, in short term studies felodipine/metoprolol had a similar (minimal) effect on QOL to enalapril monotherapy but was more cost effective.
Product Description
Felodipine, European Pharmacopoeia (EP) Reference Standard