Merck
The binding site of sodium in the gramicidin A channel.
Novartis Foundation symposium (1999-09-03)
B Roux, T B Woolf
ABSTRACT
The available information concerning the structure and location of the main binding site for sodium in the gramicidin A channel is reviewed and discussed. Results from molecular dynamics simulations using an atomic model of the channel embedded in a lipid bilayer are compared with experimental observations. The combined information from experiment and simulation suggests that the main binding sites for sodium are near the channel's mouth, approximately 9.2 A from the centre of the dimer channel, although the motion along the axis could be as large as 1 to 2 A. In the binding site, the sodium ion is lying off axis, making contact with two carbonyl oxygens and two single-file water molecules. The main channel ligand is provided by the carbonyl group of the Leu10-Trp11 peptide linkage, which exhibits the largest deflection from the ion-free channel structure.
Product Description
Gramicidin, European Pharmacopoeia (EP) Reference Standard