Merck
  • Nuclear expression of β-catenin predicts the efficacy of meloxicam treatment for patients with sporadic desmoid tumors.
Nuclear expression of β-catenin predicts the efficacy of meloxicam treatment for patients with sporadic desmoid tumors.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-01-07)
Shunsuke Hamada, Hiroshi Urakawa, Eiji Kozawa, Naohisa Futamura, Kunihiro Ikuta, Yoshie Shimoyama, Shigeo Nakamura, Naoki Ishiguro, Yoshihiro Nishida
ABSTRACT
This study aimed to determine the prevalence of β-catenin nuclear positivity as a prognostic factor in patients with desmoid tumors (DTs) treated with meloxicam, a cyclooxygenase-2 (COX-2) selective inhibitor. Between 2003 and 2012, consecutive 31 patients with extraabdominal, sporadic DTs were prospectively treated with meloxicam as a systemic medical therapy. Immunohistochemistry was performed on formalin-fixed material to quantify the nuclear expression of β-catenin and Ki-67, and cytoplasmic expression of COX-2. All clinicopathological characteristics including the intensity of immunohistochemical staining were analyzed with respect to their prognostic value for meloxicam treatment. Of the 31 patients with meloxicam treatment, there was 1 with complete remission (CR), 7 with partial remission (PR), 12 with stable disease (SD), and 11 with progressive disease (PD). Higher nuclear expression of β-catenin was significantly associated with a poor response (PD/SD) (p = 0.017). The positivity of COX-2 and Ki-67 and none of the other clinical variables were associated with prognosis. The nuclear expression of β-catenin can predict the efficacy of meloxicam treatment for patients with sporadic DTs.