Merck
  • First total synthesis of the neuronal cell protecting carbazole alkaloid carbazomadurin A by sequential transition metal-catalyzed reactions.
First total synthesis of the neuronal cell protecting carbazole alkaloid carbazomadurin A by sequential transition metal-catalyzed reactions.
Chemical communications (Cambridge, England) (2003-06-05)
Hans-Joachim Knölker, Jan Knöll
ABSTRACT
The highly oxygenated neuronal cell protecting carbazole alkaloid carbazomadurin A was synthesized in nine steps and 11% overall yield from isovanillic acid.