Merck
  • Effects of amino acids on the amidation of polyaromatic carboxylic acids by Bacillus cereus.
Effects of amino acids on the amidation of polyaromatic carboxylic acids by Bacillus cereus.
Bioscience, biotechnology, and biochemistry (2001-10-02)
R Maruyama, A Kawata, S Ono, M Nishizawa, S Ito, M Inoue
ABSTRACT
The soil bacterium Bacillus cereus Tim-r01 efficiently transformed polyaromatic carboxylic acids (PACA) such as 4-biphenylcarboxylic acid (4-BPCA), 4-biphenylacetic acid, and 4-phenoxybenzoic acid into their corresponding amides. The amidation activity was expressed at 37 degrees C (pH 7-8) in the presence of grown cells in nutrients under an aerobic atmosphere. Other strains of B. cereus, IFO 3001 and IAM 1229, also gave the amide from 4-BPCA. In phosphate-buffered saline (PBS), the addition of normal amino acids was essential, while sulfur-containing amino acids such as methionine and cysteine drastically inhibited the amidation. Tracer experiments using N-15-isoleucine and N-15-alanine showed that the nitrogen atom of the amide came from an amino group of amino acids but not from ammonia or alkylamines.