Merck
Synthesis of enantiopure 2-C-glycosyl-3-nitrochromenes.
The Journal of organic chemistry (2013-11-28)
Raquel G Soengas, Humberto Rodríguez-Solla, Artur M S Silva, Ricardo Llavona, Filipe A Almeida Paz
ABSTRACT
A novel methodology has been developed to obtain enantiopure 2-C-glycosyl-3-nitrochromenes. First, (Z)-1-bromo-1-nitroalkenes were prepared from the corresponding sugar aldehydes through a sodium iodide-catalyzed Henry reaction with bromonitromethane followed by elimination of the resulting 1-bromo-1-nitroalkan-2-ols. In the next step, reaction of the sugar-derived (Z)-1-bromo-1-nitroalkenes with o-hydroxybenzaldehydes afforded enantiopure (2S,3S,4S)-3-bromo-3,4-dihydro-4-hydroxy-3-nitro-2H-1-benzopyrans, which, upon SmI2-promoted β-elimination, yielded chiral enantiopure 2-C-glycosyl-3-nitrochromenes.