Merck
Synthesis and biological activity of novel thiazolidinediones.
Bioorganic & medicinal chemistry letters (1999-01-05)
C Prabhakar, G Madhusudhan, K Sahadev, C M Reddy, M R Sarma, G O Reddy, R Chakrabarti, C S Rao, T D Kumar, R Rajagopalan
ABSTRACT
Novel compounds having a dual pharmacophore were synthesised and evaluated for their insulin sensitiser and anti-inflammatory properties in different animal models.