Merck
Rubiadin, a new antioxidant from Rubia cordifolia.
Indian journal of biochemistry & biophysics (1997-06-01)
Y B Tripathi, M Sharma, M Manickam
ABSTRACT
Rubiadin, a dihydroxy anthraquinone, isolated from alcoholic extract of Rubia cordifolia, possesses potent antioxidant property. It prevents lipid peroxidation induced by FeSO4 and t-butylhydroperoxide (t-BHP) in a dose dependent manner. The per cent inhibition was more in the case of Fe2+ induced lipid peroxidation. The antioxidant property of the preparation has been found to be better than that of EDTA, Tris, mannitol, Vitamin E and p-benzoquinone.