Merck
[A method of increasing the effectiveness of anti-ulcer therapy].
Klinicheskaia meditsina (1989-10-01)
V G Mysh, I N Grigo'reva
ABSTRACT
Forty-three patients with duodenal ulcer were treated with almagel and gastrocepin, had four meals a day. Twenty-one patients received the drugs according to the standard schedule before meals. The test group of 22 patients took meals and drugs in turn during 24 hours with the aim of a continuous moderate reduction of acidity and activity of proteolytic enzymes. Statistical analysis of the results showed a shorter duration of the ulcer healing in the test group. This conclusion permits recommending the above novel modification of the standard treatment for use in duodenal ulcer treatment as advantageous.