Merck
  • Development of novel ferulic acid derivatives as potent histone deacetylase inhibitors.
Development of novel ferulic acid derivatives as potent histone deacetylase inhibitors.
Bioorganic & medicinal chemistry (2013-10-08)
Fang Wang, Wen Lu, Tao Zhang, Jinyun Dong, Hongping Gao, Pengfei Li, Sicen Wang, Jie Zhang
ABSTRACT
Histone deacetylase inhibitors (HDACIs) offer a promising strategy for cancer therapy. The discovery of potent ferulic acid-based HDACIs with hydroxamic acid or 2-aminobenzamide group as zinc binding group was reported. The halogeno-acetanilide was introduced as novel surface recognition moiety (SRM). The majority of title compounds displayed potent HDAC inhibitory activity. In particular, FA6 and FA16 exhibited significant enzymatic inhibitory activities, with IC50 values of 3.94 and 2.82 μM, respectively. Furthermore, these compounds showed moderate antiproliferative activity against a panel of human cancer cells. FA17 displayed promising profile as an antitumor candidate. The results indicated that these ferulic acid derivatives could serve as promising lead compounds for further optimization.