- A case of Drug Rash with Eosinophilia and Systemic Symptoms (DRESS) related to rufinamide.
A case of Drug Rash with Eosinophilia and Systemic Symptoms (DRESS) related to rufinamide.
Dermatology online journal (2013-09-12)
S Shahbaz, R K Sivamani, T Konia, B Burrall
PMID24021364
ABSTRACT
Drug Rash (or Reaction) with Eosinophilia and Systemic Symptoms (DRESS) is a potentially life-threatening hypersensitivity reaction to drugs characterized by rash, fever, lymphadenopathy, hematologic abnormalities, and involvement of internal organs. Initially coined in 1996, the term is used to refer to an idiosyncratic reaction to several drugs, the most common of which are carbamazepine, allopurinol, sulfasalazine, and phenobarbital. We report the first case of DRESS related to rufinamide in a ten year old boy with a history of a complex seizure disorder.