Merck
  • [Studies on chemical constituents from roots of Pterospermum heterophyllum].
[Studies on chemical constituents from roots of Pterospermum heterophyllum].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2008-12-18)
Yan Shi, Shuai Li, Hong-yu Li, Bao-song Cui, Yi Yuan
ABSTRACT
To study the chemical constituents from the roots of Pterospermum heterophyllum. The chemical constituents of P. heterophyllum were isolated and purified by silica gel and sephadex LH-20 column chromatography. Their structures were identified on the basis of spectroscopic data and physicochemical properties. Nine compounds were isolated and identified as taraxerol (1), betulin (2), betulinic acid (3), sumaresinolic acid (4), 2-methoxy-5-hydroxy-1, 4-naphthoquinone (5), 5, 7-dihydroxy-6, 8-dimethylchromone (6), alpha-monpalmitin (7), palmitic acid (8), beta-sitosterol (9). Compounds 2-7 were isolated from this genus for the first time.