Merck
  • Does the use of nizatidine, as a pro-kinetic agent, improve gastric emptying in patients post-oesophagectomy?
Does the use of nizatidine, as a pro-kinetic agent, improve gastric emptying in patients post-oesophagectomy?
Journal of gastrointestinal surgery : official journal of the Society for Surgery of the Alimentary Tract (2008-11-04)
Paul J Simpson, Corinne Ooi, Jennifer Chong, Andrew Smith, Andrew Baldey, Margaret Staples, Simon Woods
ABSTRACT
Delayed gastric emptying following oesophagectomy is common and can often lead to weight loss, malnutrition and a poor quality of life. Animal models have shown that nizatidine, a histamine H2-receptor antagonist, has pro-kinetic properties and can accelerate gastric emptying. Patients post-oesophagectomy require long-term acid suppression medication; if nizatidine can improve gastric emptying, it can be adopted for its dual pharmacological actions. Twenty consecutive patients were prospectively enrolled in this trial following oesophagectomy. All patients were more than 6 months post-surgery and had no evidence of recurrent cancer. A baseline nuclear medicine scan following a radiolabelled meal was conducted and then repeated after 1 week of nizatidine (150 mg bd) treatment. Quality of life and eating comfort data were collected. Oesophagectomy causes a significant delay in gastric emptying. Early satiety (80%) and reflux (65%) were the most common post-operative complaints. The percentage of food remaining in the stomach at 60 min post-meal was significantly more than normal values in both the pre- and post-nizatidine studies. There is no advantage in using nizatidine as a pro-kinetic agent. Impaired gastric emptying post-surgery causes a change in eating habits. Patients in this study did not lose a significant amount of weight despite all indicating worse eating comfort. Patients required more regular meals or snacks throughout the day and avoid foods that are difficult to swallow. It is likely that gastric motility only plays a small role in the emptying process and gravity combined with appropriate drainage procedures (pyloroplasty/pyloromyotomy) at the time of surgery are more important.