- [The deleterious effect of certain surface active agents on the ocular surface].
[The deleterious effect of certain surface active agents on the ocular surface].
Bulletin de la Societe belge d'ophtalmologie (2007-08-28)
O Ravet
PMID17718239
ABSTRACT
Most ocular solutions enter the eye through the corneal epithelial barrier. In order to pass through this barrier, these hydrosoluble drugs require to be associated with a detergent agent to increase topical efficiency. Although these agents have a preservative action, it was recently demonstrated that, after short or long term use, toxic side effects on the ocular surface will occur.