- [Medium sized rings from alprenolol and oxprenolol].
[Medium sized rings from alprenolol and oxprenolol].
Die Pharmazie (2004-11-17)
K Görlitzer, A Lorenz
PMID15544054
ABSTRACT
The N-allyl derivatives 2 formed from the beta-blockers 1, were acetylated to give the esters 5. The 1,5-benzoxazacycloundecine 6a and the 1,10,5-benzodioxazacyclododecine 7a were isolated by ring closing metathesis (RCM) of 5a and 5b, respectively, using Grubbs catalyst in the presence of titanium tetraisopropanolate. Alkaline hydrolysis yielded the alcohols 6b and 7b.