Merck
Plumieride from Allamanda cathartica as an antidermatophytic agent.
Phytotherapy research : PTR (2002-07-12)
T N Tiwari, V B Pandey, N K Dubey
ABSTRACT
Plumieride has been isolated as an active principle of the leaves of Allamanda cathartica. It showed strong fungitoxicity against some dermatophytes causing dermatomycosis to animals and human beings. It exhibited a noncytotoxic nature against a P(388) mouse leukaemia cell line.