Merck
Enzymatic hydrolysis of O-acyl esters of oxprenolol.
Acta poloniae pharmaceutica (2000-12-29)
M Nogowska
ABSTRACT
The stability of each O-acyl derivative was investigated in a solution of hydrochloric acid, artificial gastric juice, phosphate buffer at pH 7.4 and in artificial intestinal juice. A study of the relationship between the structure and enzymatic hydrolysis of the homologous series of oxprenolol esters was made.