Merck
  • Asymmetric organocatalytic Strecker-type reactions of aliphatic N,N-dialkylhydrazones.
Asymmetric organocatalytic Strecker-type reactions of aliphatic N,N-dialkylhydrazones.
Organic & biomolecular chemistry (2013-10-30)
Aurora Martínez-Muñoz, David Monge, Eloísa Martín-Zamora, Eugenia Marqués-López, Eleuterio Álvarez, Rosario Fernández, José M Lassaletta
ABSTRACT
The enantioselective organocatalytic Strecker-type reaction of aliphatic N,N-dialkylhydrazones is presented. Using trimethylsilyl cyanide (TMSCN) as the cyanide source, the reaction can be efficiently catalyzed by a tert-leucine-derived bifunctional thiourea to afford the corresponding hydrazino nitriles in good to excellent yields (50-96%) and moderate to good enantioselectivities, up to 86% ee. Further transformations of the nitrile functionality allow access to useful protected hydrazino acids and imidazolidinones. Interestingly, some of the hydrazino nitriles and their derivatives could be recrystallized in high recovery, yielding essentially pure enantiomers.