Merck
  • Prolactin is not required for the development of severe chronic experimental autoimmune encephalomyelitis.
Prolactin is not required for the development of severe chronic experimental autoimmune encephalomyelitis.
Journal of immunology (Baltimore, Md. : 1950) (2013-07-26)
Massimo Costanza, Silvia Musio, Mhamad Abou-Hamdan, Nadine Binart, Rosetta Pedotti
ABSTRACT
Predominance of multiple sclerosis (MS) in women, reductions of disease flares during pregnancy, and their increase in the postpartum period have suggested a hormonal influence on MS activity. The hormone prolactin (PRL) has long been debated as a potential immune-stimulating factor in several autoimmune disorders, including MS and its animal model experimental autoimmune encephalomyelitis (EAE). However, to date, no data clearly ascribe a pathogenic role to PRL in these diseases. Using PRL receptor-deficient (Prlr(-/-)) and PRL-deficient (Prl(-/-)) mice, we show that PRL plays a redundant role in the development of chronic EAE. In Prlr(-/-) and Prl(-/-) mice, EAE developed with a delayed onset compared with littermate control mice, but with full clinical severity. In line with the clinical outcome, T cell proliferation and production of IFN-γ, IL-17A, and IL-6 induced by myelin Ag were delayed in Prlr(-/-) and Prl(-/-) mice. Ag-specific IgG Ab responses were not affected by PRLR or PRL deficiency. We also show that mouse lymph node cells and purified CD4(+) T cells express transcript for Prlr, but not for Prl. These results reveal that PRL does not play a central role in the development of chronic EAE and optimal Th1 and Th17 responses against myelin. Moreover, they also rule out a possible contribution of PRL secreted by immune cells to the modulation of autoreactive T cell response in this model.