Merck
  • Oxidative stress decreases uptake of neutral amino acids in a human placental cell line (BeWo cells).
Oxidative stress decreases uptake of neutral amino acids in a human placental cell line (BeWo cells).
Reproductive toxicology (Elmsford, N.Y.) (2013-06-29)
João Ricardo Araújo, Ana Correia-Branco, Ana Cláudia Pereira, Maria João Pinho, Elisa Keating, Fátima Martel
ABSTRACT
Increased oxidative stress (OS) is implicated in the pathophysiology of several pregnancy disorders. We aimed to investigate the effect of tert-butylhydroperoxide (TBHP)-induced OS upon the placental transport of the neutral amino acids L-methionine (L-Met) and L-alanine (L-Ala), by using a human trophoblast cell model (BeWo cells). TBHP reduced both total and Na(+)-independent (14)C-L-Met intracellular steady-state accumulation over time (Amax), by reducing non-system L-mediated uptake - most probably system y(+) - while having no effect on system L. Moreover, TBHP reduced total (14)C-L-Ala Amax through an inhibition of system A. The effect of TBHP upon total, but not system A-mediated, (14)C-L-Ala uptake was dependent upon phosphoinositide 3-kinase (PI3K) and protein kinase C (PKC) activation, and was completely prevented by the polyphenol quercetin. In conclusion, a reduction in placental uptake of neutral amino acids may contribute to the deleterious effects of pregnancy disorders associated with OS.
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O