- Synthetic Studies Towards the Core Structure of Nakadomarin A by a Thioamide-Based Strategy.
Synthetic Studies Towards the Core Structure of Nakadomarin A by a Thioamide-Based Strategy.
European journal of organic chemistry (2014-05-16)
Jai K Chavda, Panayiotis A Procopiou, Peter N Horton, Simon J Coles, Michael J Porter
PMID24829538
ABSTRACT
The tricyclic BCD substructure of the marine natural product nakadomarin A has been synthesised. The strategy utilised a key carbon-carbon bond-forming reaction between a furan and an